2-Oxa-7-azaspiro[4.4]nonane
Supplier: Apollo Scientific
OR921290-1G OR921290-500MG
APOSOR921290-1GEA 1040 EUR
APOSOR921290-1G APOSOR921290-500MG
2-Oxa-7-azaspiro[4.4]nonane
2-Oxa-7-azaspiro[4.4]nonane
Formula: C₇H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD14581261
CAS Number: 175-97-3