1,2,3,4-Tetrahydro-1,6-naphthyridine 95%
Supplier: Apollo Scientific
Warning
OR921392-100MG OR921392-1G OR921392-250MG
APOSOR921392-100MGEA 37.7 EUR
APOSOR921392-100MG APOSOR921392-1G APOSOR921392-250MG
1,2,3,4-Tetrahydro-1,6-naphthyridine 95%
1,2,3,4-Tetrahydro-1,6-naphthyridine
Formula: C₈H₁₀N₂
MW: 134.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234763
CAS Number: 13623-84-2