1H-Pyrrolo[3,2-b]pyridine-3-carbonitrile 95%
Supplier: Apollo Scientific
OR921537-1G OR921537-250MG
APOSOR921537-1GEA 142 EUR
APOSOR921537-1G APOSOR921537-250MG
1H-Pyrrolo[3,2-b]pyridine-3-carbonitrile 95%
1H-Pyrrolo[3,2-b]pyridine-3-carbonitrile
Formula: C₈H₅N₃
MW: 143.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12963980
CAS Number: 1196151-62-8