2-Bromo-1H-pyrrolo[2,3-c]pyridine ≥98%
Supplier: Apollo Scientific
OR921599-100MG OR921599-1G OR921599-250MG
APOSOR921599-100MGEA 409 EUR
APOSOR921599-100MG APOSOR921599-1G APOSOR921599-250MG
2-Bromo-1H-pyrrolo[2,3-c]pyridine ≥98%
2-Bromo-1H-pyrrolo[2,3-c]pyridine
Formula: C₇H₅BrN₂
MW: 197.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738911
CAS Number: 885272-38-8