2,2-Pentamethylene-1,3-dioxolane 95%
Supplier: Apollo Scientific
Synonyms: 1,4-Dioxaspiro[4.5]decane
OR921676-1G OR921676-5G
APOSOR921676-1GEA 42.2 EUR
APOSOR921676-1G APOSOR921676-5G
2,2-Pentamethylene-1,3-dioxolane 95%
2,2-Pentamethylene-1,3-dioxolane
Formula: C₈H₁₄O₂
MW: 142.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005409
CAS Number: 177-10-6