Methyl 5-(chlorosulphonyl)pentanoate 95%
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 5-(Chlorosulphonyl)pentanoate
OR921979-100MG OR921979-1G OR921979-250MG
APOSOR921979-100MGEA 98.6 EUR
APOSOR921979-100MG APOSOR921979-1G APOSOR921979-250MG
Methyl 5-(chlorosulphonyl)pentanoate 95%
Methyl 5-(chlorosulphonyl)pentanoate
Formula: C₆H₁₁ClO₄S
MW: 214.667 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22574974
CAS Number: 1408058-14-9
UN: 3265
ADR: 8,III