3-Methylpiperidin-4-ol hydrochloride 95%
Supplier: Apollo Scientific
OR922447-1G OR922447-5G
APOSOR922447-1GEA 207 EUR
APOSOR922447-1G APOSOR922447-5G
3-Methylpiperidin-4-ol hydrochloride 95%
3-Methylpiperidin-4-ol hydrochloride
Formula: C₆H₁₃NO·ClH
MW: 151.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11506421
CAS Number: 1185293-84-8