(4-Ethoxy-phenyl)-oxo-acetonitrile 95%
Supplier: Apollo Scientific
OR922540-1G OR922540-250MG
APOSOR922540-1GEA 864 EUR
APOSOR922540-1G APOSOR922540-250MG
(4-Ethoxy-phenyl)-oxo-acetonitrile 95%
(4-Ethoxy-phenyl)-oxo-acetonitrile
Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658436
CAS Number: 62869-42-5