3,3-Dimethylpyrrolidine hydrochloride 95%
Supplier: Apollo Scientific
OR922548-100MG OR922548-1G OR922548-250MG
APOSOR922548-100MGEA 52.2 EUR
APOSOR922548-100MG APOSOR922548-1G APOSOR922548-250MG
3,3-Dimethylpyrrolidine hydrochloride 95%
3,3-Dimethylpyrrolidine hydrochloride
Formula: C₆H₁₃N·ClH
MW: 135.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849606
CAS Number: 3437-30-7