2,7-Diazaspiro[4.4]nonan-1-one hydrochloride 97%
Supplier: Apollo Scientific
OR922643-1G OR922643-500MG
APOSOR922643-1GEA 690 EUR
APOSOR922643-1G APOSOR922643-500MG
2,7-Diazaspiro[4.4]nonan-1-one hydrochloride 97%
2,7-Diazaspiro[4.4]nonan-1-one hydrochloride
Formula: C₇H₁₂N₂O·ClH
MW: 176.644 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10700079
CAS Number: 1203682-17-0