4-(2-Hydroxyethoxy)-3,5-dimethylbenzaldehyde 95%
Supplier: Apollo Scientific
OR923041-1G OR923041-250MG
APOSOR923041-1GEA 270 EUR
APOSOR923041-1G APOSOR923041-250MG
4-(2-Hydroxyethoxy)-3,5-dimethylbenzaldehyde 95%
4-(2-Hydroxyethoxy)-3,5-dimethylbenzaldehyde
Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11188381
CAS Number: 1039948-89-4