3-Acetyl-2,6-dimethoxypyridine 97%
Supplier: Apollo Scientific
OR923223-10G OR923223-1G OR923223-5G
APOSOR923223-10GEA 538 EUR
APOSOR923223-10G APOSOR923223-1G APOSOR923223-5G
3-Acetyl-2,6-dimethoxypyridine 97%
3-Acetyl-2,6-dimethoxypyridine
Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784407
CAS Number: 870703-62-1