6-(Morpholin-4-yl)pyridine-2-carboxaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(6-Formylpyridin-2-yl)morpholine, 2-Formyl-6-(morpholin-4-yl)pyridine
OR9234-1G OR9234-250MG
APOSOR9234-1GEA 382 EUR
APOSOR9234-1G APOSOR9234-250MG
6-(Morpholin-4-yl)pyridine-2-carboxaldehyde 97%
6-(Morpholin-4-yl)pyridine-2-carboxaldehyde
Formula: C₁₀H₁₂N₂O₂
MW: 192.214 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD08271893
CAS Number: 857283-88-6