cis-3-Hydroxy-3-methylcyclobutanecarboxylic acid
Supplier: Apollo Scientific
OR923565-1G OR923565-5G
APOSOR923565-1GEA 543 EUR
APOSOR923565-1G APOSOR923565-5G
cis-3-Hydroxy-3-methylcyclobutanecarboxylic acid
cis-3-Hydroxy-3-methylcyclobutanecarboxylic acid
Formula: C₆H₁₀O₃
MW: 130.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106032
CAS Number: 1384855-41-7