8-Oxa-2-azaspiro[4.5]decan-3-one
Supplier: Apollo Scientific
OR923853-1G OR923853-5G
APOSOR923853-1GEA 71.2 EUR
APOSOR923853-1G APOSOR923853-5G
8-Oxa-2-azaspiro[4.5]decan-3-one
8-Oxa-2-azaspiro[4.5]decan-3-one
Formula: C₈H₁₃NO₂
MW: 155.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13180589
CAS Number: 194862-84-5