4-Bromo-1H-pyrazolo[3,4-d]pyrimidine 95%
Supplier: Apollo Scientific
OR923883-1G OR923883-250MG
APOSOR923883-1GEA 806 EUR
APOSOR923883-1G APOSOR923883-250MG
4-Bromo-1H-pyrazolo[3,4-d]pyrimidine 95%
4-Bromo-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₅H₃BrN₄
MW: 199.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040189
CAS Number: 30129-51-2