Pyrido[3,2-d]pyrimidin-4(3H)-one 96%
Supplier: Apollo Scientific
OR923956-1G
APOSOR923956-1GEA 94.4 EUR
APOSOR923956-1G
Pyrido[3,2-d]pyrimidin-4(3H)-one 96%
Pyrido[3,2-d]pyrimidin-4(3H)-one
Formula: C₇H₅N₃O
MW: 147.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02645992
CAS Number: 37538-67-3