4-Methoxy-7h-pyrrolo[2,3-d]pyrimidine 98%
Supplier: Apollo Scientific
Synonyms: 4-Methoxy-7H-pyrrolo[2,3-d]pyrimidine
OR924446-1G OR924446-5G
APOSOR924446-1GEA 148 EUR
APOSOR924446-1G APOSOR924446-5G
4-Methoxy-7h-pyrrolo[2,3-d]pyrimidine 98%
4-Methoxy-7h-pyrrolo[2,3-d]pyrimidine
Formula: C₇H₇N₃O
MW: 149.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127852
CAS Number: 4786-76-9