1-(4-Methylthiazol-2-yl)ethanone 95%
Supplier: Apollo Scientific
OR924451-1G OR924451-5G
APOSOR924451-1GEA 95.7 EUR
APOSOR924451-1G APOSOR924451-5G
1-(4-Methylthiazol-2-yl)ethanone 95%
1-(4-Methylthiazol-2-yl)ethanone
Formula: C₆H₇NOS
MW: 141.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369793
CAS Number: 7533-07-5