1H,4H-Pyrazolo[1,5-a][1,3,5]triazin-4-one 95%
Supplier: Apollo Scientific
OR924884-1G OR924884-250MG
APOSOR924884-1GEA 137 EUR
APOSOR924884-1G APOSOR924884-250MG
1H,4H-Pyrazolo[1,5-a][1,3,5]triazin-4-one 95%
1H,4H-Pyrazolo[1,5-a][1,3,5]triazin-4-one
Formula: C₅H₄N₄O
MW: 136.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19227971
CAS Number: 54346-27-9