(3R)-1-Isopropylpyrrolidin-3-amine 95%
Supplier: Apollo Scientific
OR924898-1G OR924898-250MG
APOSOR924898-1GEA 841 EUR
APOSOR924898-1G APOSOR924898-250MG
(3R)-1-Isopropylpyrrolidin-3-amine 95%
(3R)-1-Isopropylpyrrolidin-3-amine
Formula: C₇H₁₆N₂
MW: 128.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264184
CAS Number: 935534-43-3