6-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine 95%
Supplier: Apollo Scientific
OR924956-1G OR924956-250MG
APOSOR924956-1GEA 806 EUR
APOSOR924956-1G APOSOR924956-250MG
6-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine 95%
6-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine
Formula: C₈H₁₁N₃
MW: 149.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22565616
CAS Number: 66521-83-3