4-Chloro-6-methylthieno[2,3-d]pyrimidine 95%
Supplier: Apollo Scientific
OR925626-1G OR925626-250MG
APOSOR925626-1GEA 538 EUR
APOSOR925626-1G APOSOR925626-250MG
4-Chloro-6-methylthieno[2,3-d]pyrimidine 95%
4-Chloro-6-methylthieno[2,3-d]pyrimidine
Formula: C₇H₅ClN₂S
MW: 184.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03030440
CAS Number: 106691-21-8