2-Methoxy-6-methyl-4(1H)-pyrimidinone 95%
Supplier: Apollo Scientific
Synonyms: 2-Methoxy-6-methylpyrimidin-4(1H)-one
OR925628-1G
APOSOR925628-1GEA 46.4 EUR
APOSOR925628-1G
2-Methoxy-6-methyl-4(1H)-pyrimidinone 95%
2-Methoxy-6-methyl-4(1H)-pyrimidinone
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743870
CAS Number: 55996-28-6