1-(2-Chloro-1H-indol-3-yl)-ethanone 95%
Supplier: Apollo Scientific
OR925896-1G OR925896-250MG
APOSOR925896-1GEA 683 EUR
APOSOR925896-1G APOSOR925896-250MG
1-(2-Chloro-1H-indol-3-yl)-ethanone 95%
1-(2-Chloro-1H-indol-3-yl)-ethanone
Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02956036
CAS Number: 65287-74-3