6-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine 95%
Supplier: Apollo Scientific
OR926202-1G
OR926202-250MG
APOSOR926202-1GEA
719
EUR
APOSOR926202-1G
APOSOR926202-250MG
6-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine 95%
6-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula:
C₇H₈N₄ MW: 148.165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846607 CAS Number: 1239647-61-0 |