2-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine hydrochloride 95%
Supplier: Apollo Scientific
OR926218-1G OR926218-250MG
APOSOR926218-1GEA 719 EUR
APOSOR926218-1G APOSOR926218-250MG
2-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine hydrochloride 95%
2-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine hydrochloride
Formula: C₈H₁₁N₃·ClH
MW: 185.654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11043222
CAS Number: 210538-73-1