6,7-Dihydro-5h-cyclopenta[b]pyridin-7-ol 95%
Supplier: Apollo Scientific
Synonyms: 6,7-Dihydro-5H-cyclopenta[b]pyridin-7-ol
OR926347-1G OR926347-250MG OR926347-5G
APOSOR926347-1GEA 115 EUR
APOSOR926347-1G APOSOR926347-250MG APOSOR926347-5G
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-ol 95%
6,7-Dihydro-5h-cyclopenta[b]pyridin-7-ol
Formula: C₈H₉NO
MW: 135.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778342
CAS Number: 41598-71-4