Morpholin-3-ylmethanol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxymethylmorpholine
OR926444-1G OR926444-5G
APOSOR926444-1GEA 299 EUR
APOSOR926444-1G APOSOR926444-5G
Morpholin-3-ylmethanol 96%
Morpholin-3-ylmethanol
Formula: C₅H₁₁NO₂
MW: 117.146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038650
CAS Number: 106910-83-2