Dimethyl 1H-pyrrole-2,4-dicarboxylate 95%
Supplier: Apollo Scientific
OR926541-1G OR926541-250MG
APOSOR926541-1GEA 53.8 EUR
APOSOR926541-1G APOSOR926541-250MG
Dimethyl 1H-pyrrole-2,4-dicarboxylate 95%
Dimethyl 1H-pyrrole-2,4-dicarboxylate
Formula: C₈H₉NO₄
MW: 183.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179576
CAS Number: 2818-07-7