N-(4-tert-Butyl-phenyl)-formamide 95%
Supplier: Apollo Scientific
OR926571-1G OR926571-250MG
APOSOR926571-1GEA 574 EUR
APOSOR926571-1G APOSOR926571-250MG
N-(4-tert-Butyl-phenyl)-formamide 95%
N-(4-tert-Butyl-phenyl)-formamide
Formula: C₁₁H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD08457067
CAS Number: 63429-97-0