3-Methylcyclobutane-1-carboxylic acid
Supplier: Apollo Scientific
Synonyms: 3-Methylcyclobutanecarboxylic acid
OR926901-1G OR926901-25G OR926901-5G
APOSOR926901-1GEA 105 EUR
APOSOR926901-1G APOSOR926901-25G APOSOR926901-5G
3-Methylcyclobutane-1-carboxylic acid
3-Methylcyclobutane-1-carboxylic acid
Formula: C₆H₁₀O₂
MW: 114.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01734189
CAS Number: 57252-83-2