Pyrazolo[1,5-a]pyrimidine-5,7-diol 96%
Supplier: Apollo Scientific
Synonyms: 7-Hydroxypyrazolo[1,5-a]pyrimidin-5(4H)-one
OR927008-10G OR927008-1G OR927008-5G
APOSOR927008-10GEA 474 EUR
APOSOR927008-10G APOSOR927008-1G APOSOR927008-5G
Pyrazolo[1,5-a]pyrimidine-5,7-diol 96%
Pyrazolo[1,5-a]pyrimidine-5,7-diol
Formula: C₆H₅N₃O₂
MW: 151.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02091625
CAS Number: 57489-70-0