3-Ethoxy-5-methyl-1H-pyrazole 98%
Supplier: Apollo Scientific
OR927527-1G OR927527-250MG
APOSOR927527-1GEA 406 EUR
APOSOR927527-1G APOSOR927527-250MG
3-Ethoxy-5-methyl-1H-pyrazole 98%
3-Ethoxy-5-methyl-1H-pyrazole
Formula: C₆H₁₀N₂O
MW: 126.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234240
CAS Number: 3201-21-6