2-Chloro-4,6-dimethylnicotinic acid 97%
Supplier: Apollo Scientific
OR927545-1G OR927545-5G
APOSOR927545-1GEA 49.3 EUR
APOSOR927545-1G APOSOR927545-5G
2-Chloro-4,6-dimethylnicotinic acid 97%
2-Chloro-4,6-dimethylnicotinic acid
Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02110082
CAS Number: 66662-48-4