(S)-Chroman-3-amine hydrochloride 95%
Supplier: Apollo Scientific
Synonyms: (S)-3-Aminochromane Hydrochloride
OR927573-1G OR927573-250MG
APOSOR927573-1GEA 232 EUR
APOSOR927573-1G APOSOR927573-250MG
(S)-Chroman-3-amine hydrochloride 95%
(S)-Chroman-3-amine hydrochloride
Formula: C₉H₁₁NO·ClH
MW: 185.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22574992
CAS Number: 211506-60-4