3-Methoxy-4-(1,3-oxazol-5-yl)aniline 98%
Supplier: Apollo Scientific
Synonyms: 3-Methoxy-4-(oxazol-5-yl)aniline
OR927626-1G OR927626-250MG
APOSOR927626-1GEA 97.3 EUR
APOSOR927626-1G APOSOR927626-250MG
3-Methoxy-4-(1,3-oxazol-5-yl)aniline 98%
3-Methoxy-4-(1,3-oxazol-5-yl)aniline
Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844645
CAS Number: 198821-79-3