N,N'-Dimethyl-1,4-butanediamine 95%
Supplier: Apollo Scientific
Synonyms: N1,N4-Dimethylbutane-1,4-diamine
OR927887-5G
APOSOR927887-5GEA 79.9 EUR
APOSOR927887-5G
N,N'-Dimethyl-1,4-butanediamine 95%
N,N'-Dimethyl-1,4-butanediamine
Formula: C₆H₁₆N₂
MW: 116.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039799
CAS Number: 16011-97-5