8-Methoxyquinazolin-4-ol 95%
Supplier: Apollo Scientific
OR927982-1G OR927982-250MG
APOSOR927982-1GEA 226 EUR
APOSOR927982-1G APOSOR927982-250MG
8-Methoxyquinazolin-4-ol 95%
8-Methoxyquinazolin-4-ol
Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527388
CAS Number: 16064-27-0