4-Chloro-2-methylpyrimidine-5-carbonitrile 98%
Supplier: Apollo Scientific
Warning
OR928159-1G OR928159-5G
APOSOR928159-1GEA 172 EUR
APOSOR928159-1G APOSOR928159-5G
4-Chloro-2-methylpyrimidine-5-carbonitrile 98%
4-Chloro-2-methylpyrimidine-5-carbonitrile
Formula: C₆H₄ClN₃
MW: 153.569 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193459
CAS Number: 38875-74-0