5-Chlorothiophene-3-carboxaldehyde 95%
Supplier: Apollo Scientific
Synonyms: 5-Chlorothiophene-3-carbaldehyde
OR928325-1G OR928325-250MG
APOSOR928325-1GEA 473 EUR
APOSOR928325-1G APOSOR928325-250MG
5-Chlorothiophene-3-carboxaldehyde 95%
5-Chlorothiophene-3-carboxaldehyde
Formula: C₅H₃ClOS
MW: 146.595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17016115
CAS Number: 36155-85-8