(R)-(-)-1-Benzyloxy-2-propanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-1-(Benzyloxy)propan-2-ol
OR928945-1G OR928945-5G
APOSOR928945-1GEA 52.2 EUR
APOSOR928945-1G APOSOR928945-5G
(R)-(-)-1-Benzyloxy-2-propanol 95%
(R)-(-)-1-Benzyloxy-2-propanol
Formula: C₁₀H₁₄O₂
Density: 1.027 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367007
CAS Number: 89401-28-5