7-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine, tech
Supplier: Apollo Scientific
Warning
OR929053-1G OR929053-500MG
APOSOR929053-1GEA 114 EUR
APOSOR929053-1G APOSOR929053-500MG
7-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine, tech
7-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula: C₇H₈N₄
MW: 148.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846604
CAS Number: 1239648-57-7