5-(Tetrahydro-2H-pyran-2-yl)-2-furaldehyde 95%
Supplier: Apollo Scientific
OR929745-1G OR929745-5G
APOSOR929745-1GEA 168 EUR
APOSOR929745-1G APOSOR929745-5G
5-(Tetrahydro-2H-pyran-2-yl)-2-furaldehyde 95%
5-(Tetrahydro-2H-pyran-2-yl)-2-furaldehyde
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059921
CAS Number: 915924-73-1