1-Piperidin-2-yl-ethanone 95%
Supplier: Apollo Scientific
OR930107-100MG OR930107-1G OR930107-250MG
APOSOR930107-100MGEA 161 EUR
APOSOR930107-100MG APOSOR930107-1G APOSOR930107-250MG
1-Piperidin-2-yl-ethanone 95%
1-Piperidin-2-yl-ethanone
Formula: C₇H₁₃NO
MW: 127.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658295
CAS Number: 97073-22-8