1-(1,5-Dimethyl-1H-pyrazol-4-yl)ethanamine 95%
Supplier: Apollo Scientific
OR930112-1G OR930112-250MG
APOSOR930112-1GEA 647 EUR
APOSOR930112-1G APOSOR930112-250MG
1-(1,5-Dimethyl-1H-pyrazol-4-yl)ethanamine 95%
1-(1,5-Dimethyl-1H-pyrazol-4-yl)ethanamine
Formula: C₇H₁₃N₃
MW: 139.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03422568
CAS Number: 936939-85-4