1-(2-Chloro-6-methoxyphenyl)ethanone 95%

Supplier: Apollo Scientific

OR930146-1G OR930146-250MG
APOSOR930146-1GEA 75.7 EUR
APOSOR930146-1G APOSOR930146-250MG
1-(2-Chloro-6-methoxyphenyl)ethanone 95%
1-(2-Chloro-6-methoxyphenyl)ethanone
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24847834
CAS Number: 881883-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR