2-(4-Methoxy-3-nitrophenyl)acetonitrile 95%
Supplier: Apollo Scientific
OR930248-1G OR930248-250MG
APOSOR930248-1GEA 502 EUR
APOSOR930248-1G APOSOR930248-250MG
2-(4-Methoxy-3-nitrophenyl)acetonitrile 95%
2-(4-Methoxy-3-nitrophenyl)acetonitrile
Formula: C₉H₈N₂O₃
MW: 192.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11118823
CAS Number: 56245-07-9