1-(4-Chloro-7h-pyrrolo[2,3-d]pyrimidin-5-yl)ethanone 97%
Supplier: Apollo Scientific
Synonyms: 5-Acetyl-4-Chloro-7H-pyrrolo[2,3-d]pyrimidine
OR930287-1G OR930287-5G
APOSOR930287-1GEA 97.3 EUR
APOSOR930287-1G APOSOR930287-5G
1-(4-Chloro-7h-pyrrolo[2,3-d]pyrimidin-5-yl)ethanone 97%
1-(4-Chloro-7h-pyrrolo[2,3-d]pyrimidin-5-yl)ethanone
Formula: C₈H₆ClN₃O
MW: 195.606 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22205848
CAS Number: 1363381-59-2