6-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine 96%
Supplier: Apollo Scientific
OR930295-1G OR930295-250MG
APOSOR930295-1GEA 451 EUR
APOSOR930295-1G APOSOR930295-250MG
6-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine 96%
6-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formula: C₆H₅ClN₄
MW: 168.584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846610
CAS Number: 1239647-60-9